A0417
Angiotensinogen Fragment 11-14
≥97% (HPLC)
Synonym(s):
Val-Ile-His-Asn
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C21H35N7O6
CAS Number:
Molecular Weight:
481.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
Assay
≥97% (HPLC)
storage temp.
−20°C
SMILES string
CCC(C)C(NC(=O)C(N)C(C)C)C(=O)NC(Cc1cnc[nH]1)C(=O)NC(CC(N)=O)C(O)=O
Other Notes
Product of the action of human kidney renin on human angiotensinogen.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
M Poe et al.
Analytical biochemistry, 140(2), 459-467 (1984-08-01)
A synthetic tetradecapeptide, H-Asp-Arg-Val-Tyr-Ile-His-Pro-Phe-His-Leu-Val-Ile-His-Ser-OH, which corresponds to the 13 amino terminal residues of human angiotensinogen plus a carboxy terminal serine to replace a suggested site of carbohydrate attachment, has been shown to be a good substrate for human kidney renin.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service