Skip to Content
Merck
All Photos(1)

Key Documents

A0205

Sigma-Aldrich

Ala-Arg-Pro-Ala-D-Lys amide trifluoroacetate salt

≥97% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H44N10O5
CAS Number:
Molecular Weight:
540.66
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

≥97% (HPLC)

storage temp.

−20°C

SMILES string

CC(N)C(=O)NC(CCCNC(N)=N)C(=O)N1CCCC1C(=O)NC(C)C(=O)NC(CCCCN)C(N)=O

Biochem/physiol Actions

A potent coronary vasodilator.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T G Saldeen et al.
Journal of cardiovascular pharmacology, 23(1), 103-106 (1994-01-01)
Peptide 6A, a degradation of B beta chain of fibrinogen, is known to increase coronary and femoral blood flow. In this study, we examined the effects of peptide 32, an analogue of peptide 6A on coronary blood flow in anesthetized

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service