Key Documents
G6897
Gradient-forming devices
total capacity 50 mL
About This Item
UNSPSC Code:
41100000
Pricing and availability is not currently available.
total capacity
50 mL
General description
Constructed of two equal-diameter heavy-walled acrylic cylinders secured to an acrylic base. PTFE stopcock between cylinders is closed for filling, then opened. Exit port from mixing chamber is controlled by addition of hose and pinch clamp. Geometry permits use of magnetic stirring bar in mixing chamber.
Choose from one of the most recent versions:
Certificates of Analysis (COA)