Skip to Content
Merck

93987

myo-Inositol 1,2,3,4,6-pentakis-phosphate

≥85% (HPLC)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H17O21P5
CAS Number:
Molecular Weight:
580.06
PubChem Substance ID:
UNSPSC Code:
12352202
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥85% (HPLC)

form

paste

storage temp.

−20°C

SMILES string

O[C@@H]1[C@@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@H]1OP(O)(O)=O

InChI

1S/C6H17O21P5/c7-1-2(23-28(8,9)10)4(25-30(14,15)16)6(27-32(20,21)22)5(26-31(17,18)19)3(1)24-29(11,12)13/h1-7H,(H2,8,9,10)(H2,11,12,13)(H2,14,15,16)(H2,17,18,19)(H2,20,21,22)/t1-,2-,3+,4+,5-,6+

InChI key

CTPQAXVNYGZUAJ-QWBQGLJISA-N

Biochem/physiol Actions

Intermediate in inositol phosphate metabolism. Inositides have an important impact on diverse areas of cellular regulation.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library