Skip to Content
Merck

92606

Atto Oxa12

BioReagent, suitable for fluorescence

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

form

solid

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 663 nm; λem 684 nm

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto Oxa12 is a red-emitting label and a lipophilic variety of Atto 655. Characteristic features of the label are strong absorption, good fluorescence quantum yield, very little triplet formation, excellent thermal and photo-stability, and outstanding ozone resistance.Atto Oxa12 is a cationic dye (charge +1). Absorption and fluorescence are pH independent in the range pH 2 to 11, used in typical applications. Atto Oxa12 shows excellent solubility in organic solvents like DMF, DMSO or acetonitrile.


Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library