Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C22H25N5O4S
CAS Number:
Molecular Weight:
455.53
NACRES:
NA.32
PubChem Substance ID:
UNSPSC Code:
12352108
MDL number:
form
powder
fluorescence
λex 586 nm; λem 735 nm in ethanol
storage temp.
−20°C
SMILES string
CCN(CC)c1ccc2cc(\C=C\c3cc[n+](CCCN=[N+]=[N-])cc3S([O-])(=O)=O)oc2c1
InChI
1S/C22H25N5O4S/c1-3-27(4-2)19-8-6-18-14-20(31-21(18)15-19)9-7-17-10-13-26(12-5-11-24-25-23)16-22(17)32(28,29)30/h6-10,13-16H,3-5,11-12H2,1-2H3
InChI key
CGMCCNZFQJOMRA-UHFFFAOYSA-N
General description
The MegaStokes dyes for „click" biological labeling, licensed by LuminoChem are virtually covering the whole visible spectrum reaching the infra-red regime. Besides dyes that are being capable of participating in classical copper catalyzed 1,3-dipolar cycloaddition reaction with the counterparting function, we also provide dyes containing cyclooctyne moiety an alkyne derivative that enables copper free clicking to azides.
These dyes are noteworthy for their large Stokes-shift (120 or 125 nm). Dyes for this kind are exceptionally useful in fluorescence resonance electron transfer (FRET) applications as they do not interfere with the spectral bands of the second fluorophore a common problem in FRET technology. Azido sugars have been used for click-labeling of surface glycoproteins, recently. We have adapted this system to demonstrate the ability of our dyes to undergo bioorthogonal labeling reaction, efficiently. Prior to labeling, Chinese hamster ovary (CHO) cells were incubated with these MegaStockesdyes to test the possible cytostatic effects of these dyes. Experimental results have shown that neither dye is toxic up to 50 μM concentration.
These dyes are noteworthy for their large Stokes-shift (120 or 125 nm). Dyes for this kind are exceptionally useful in fluorescence resonance electron transfer (FRET) applications as they do not interfere with the spectral bands of the second fluorophore a common problem in FRET technology. Azido sugars have been used for click-labeling of surface glycoproteins, recently. We have adapted this system to demonstrate the ability of our dyes to undergo bioorthogonal labeling reaction, efficiently. Prior to labeling, Chinese hamster ovary (CHO) cells were incubated with these MegaStockesdyes to test the possible cytostatic effects of these dyes. Experimental results have shown that neither dye is toxic up to 50 μM concentration.
Packaging
Bottomless glass bottle. Contents are inside inserted fused cone.
Storage Class
13 - Non Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 87564-10MG | 04061833248652 |