Skip to Content
Merck

Skip To

82178

Sodium chloride 0.1M – Sodium acetate pH 4.6; 0.1M – 2-Propanol 12% (v/v) solution

Synonym(s):

Crystallization solution 4/Kit-No 73513

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12352200


form

solution

Quality Segment

pH

4.6±0.2 ( in neat)

density

0.99 g/mL at 20 °C

storage temp.

2-8°C



pictograms

FlameExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3

Storage Class

3 - Flammable liquids

wgk

WGK 1

flash_point_f

93.2 °F - closed cup

flash_point_c

34 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters