Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.32
UNSPSC Code:
12352108
Form:
powder
form
powder
manufacturer/tradename
ATTO-TEC GmbH
fluorescence
λex 601 nm; λem 627 nm in PBS
suitability
suitable for fluorescence
storage temp.
−20°C
Application
Fluorescent labeled ovalbumin is used as endocytic tracer.
Analysis Note
unconjugated dye ≤5% of total fluorescence
Legal Information
This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 76503-2MG | 04061832533148 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service