Skip to Content
Merck

76141

Atto Rho13 Biotin

BioReagent, suitable for fluorescence

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

form

solid

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 600 nm; λem 625 nm

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto Rho13 Biotin is a new rhodamine dye for application in life sciences, e.g. labeling of DNA, RNA or proteins. The label shows strong absorption, high fluorescence quantum yield, and high photostability. After coupling to a substrate Atto Rho13 Biotin carries a net electrical charge of +1.
The dye is moderately hydrophilic. Biotin conjugates can be used in applications like ELISA or immunohistochemistry, in situ-hybridization, flow cytometry and others, to identify streptavidin, avidin or extravidin-conjugates.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library