Skip to Content

74719

Octaethylene glycol monotetradecyl ether

≥99.0% (TLC)

Synonym(s):

C14E8, Polyoxyethylene (8) myristyl ether, Tetradecyl octaethylene glycol ether, Tetradecyloctaglycol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
CH3(CH2)13(OCH2CH2)8OH
CAS Number:
Molecular Weight:
566.81
PubChem Substance ID:
UNSPSC Code:
12161900
Beilstein/REAXYS Number:
5996901
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

non-ionic

assay

≥99.0% (TLC)

mol wt

566.81 g/mol

technique(s)

protein quantification: suitable

mp

38-39 °C

cation traces

Ca: ≤50 mg/kg, Cd: ≤50 mg/kg, Co: ≤50 mg/kg, Cr: ≤50 mg/kg, Cu: ≤50 mg/kg, Fe: ≤50 mg/kg, K: ≤50 mg/kg, Mg: ≤50 mg/kg, Mn: ≤50 mg/kg, Na: ≤50 mg/kg, Ni: ≤50 mg/kg, Pb: ≤50 mg/kg, Zn: ≤50 mg/kg

SMILES string

CCCCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C30H62O9/c1-2-3-4-5-6-7-8-9-10-11-12-13-15-32-17-19-34-21-23-36-25-27-38-29-30-39-28-26-37-24-22-35-20-18-33-16-14-31/h31H,2-30H2,1H3

InChI key

NHHAZFYVKWSFIR-UHFFFAOYSA-N



Storage Class

13 - Non Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library