Skip to Content
Merck
All Photos(1)

Key Documents

74565

Sigma-Aldrich

Eriodictyol

≥95.0% (HPLC)

Synonym(s):

(S)-3′,4′,5,7-Tetrahydroxyflavanone, Eriodictiol, Huazhongilexone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H12O6
CAS Number:
Molecular Weight:
288.25
Beilstein:
5104930
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥95.0% (HPLC)

form

powder

storage temp.

2-8°C

SMILES string

Oc1cc(O)c2C(=O)C[C@H](Oc2c1)c3ccc(O)c(O)c3

InChI

1S/C15H12O6/c16-8-4-11(19)15-12(20)6-13(21-14(15)5-8)7-1-2-9(17)10(18)3-7/h1-5,13,16-19H,6H2/t13-/m0/s1

InChI key

SBHXYTNGIZCORC-ZDUSSCGKSA-N

Gene Information

mouse ... Hexa(15211)

Biochem/physiol Actions

The flavanone eriodictyol is a metabolite in the flavonoid biosynthesis pathway. Eriodictyol has anti-inflammatory and antioxidant activities[1] and taste-modifying properties; it is extracted from plants such as Eriodictyon californicum[2], the twigs of Millettia duchesnei,[3] in Eupatorium arnottianum,[4] lemons and as glycoside in rose hips (Rosa canina).[5]

replaced by

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Erlend Hvattum
Rapid communications in mass spectrometry : RCM, 16(7), 655-662 (2002-03-29)
Liquid chromatography coupled with negative and positive electrospray ionisation (ESI) tandem mass spectrometry (MS/MS) and diode-array detection (DAD) was used for determination of phenols in rose hip (Rosa canina) extract. ESI mass spectra of the chromatographically separated phenols gave the
Manar M Salem et al.
Journal of natural products, 68(1), 108-111 (2005-02-01)
Bioactivity-guided fractionation of the methanolic extract of Psorothamnus polydenius yielded the new chalcone 2,2',4'-trihydroxy-6'-methoxy-3',5'-dimethylchalcone (2), together with six other known compounds, 2',4'-dihydroxy-6'-methoxy-3',5'-dimethylchalcone (1), dalrubone (3), demethoxymatteucinol (4), eriodictyol (5), and photodalrubone (6a and 6b). This is the first report of
François Ngandeu et al.
Phytochemistry, 69(1), 258-263 (2007-07-21)
Three prenylated rotenoids, elliptol, 12-deoxo-12alpha-methoxyelliptone and 6-methoxy-6a,12a-dehydrodeguelin were isolated from the twigs of Millettia duchesnei, together with the known compounds, 6a,12a-dehydrodeguelin, 6-hydroxy-6a,12a-dehydrodeguelin, 6-oxo-6a,12a-dehydrodeguelin, elliptone, 12a-hydroxyelliptone and eriodictyol. Their structures were elucidated on the basis of spectral data and comparison with
B A 'T Hart et al.
Chemico-biological interactions, 73(2-3), 323-335 (1990-01-01)
The mechanism by which (a panel of) flanonoids inhibit the production of luminol-dependent chemiluminescence (CLlum) by activated human neutrophils is subject to this study. CLlum is frequently used as a bio-assay to quantify the effect of xenobiotics on the production
Defang Li et al.
Frontiers in pharmacology, 9, 33-33 (2018-02-15)
Myocardial ischemia-reperfusion (I/R) injury remains the leading risk factor of disability and mortality worldwide. In this study, the myocardial protective effect of eriodictyol (EDT) and the underlying mechanism in an

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service