73938
Fluorescent IEF-Marker pI 6.7 solution
1 mg/mL in 10 mM HCl
Synonym(s):
IEF-Marker fluorescent pI 6.7
About This Item
form
liquid
mol wt
Mw 379.25 g/mol
concentration
1 mg/mL in 10 mM HCl
fluorescence
λex 394 nm; λem 505 nm (in 0.1 M phosphate pH 6.7)
application(s)
cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care
storage temp.
−20°C
General description
Application
Reconstitution
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Information on Isoelectric Focusing including what it is and how it is used. In order to ensure the high performance of analysis, isoelectric point (pI) standards are needed.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service