Skip to Content
Merck
All Photos(1)

Key Documents

69948

Sigma-Aldrich

MOPS

BioUltra, ≥99.5% (T)

Synonym(s):

3-(N-Morpholino)propanesulfonic acid, 4-Morpholinepropanesulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H15NO4S
CAS Number:
Molecular Weight:
209.26
Beilstein:
1106776
EC Number:
MDL number:
UNSPSC Code:
12161700
PubChem Substance ID:

product line

BioUltra

Assay

≥99.5% (T)

form

powder or crystals

impurities

insoluble matter, passes filter test

ign. residue

≤0.1% (as SO4)

loss

≤1% loss on drying, 110 °C

pH

2.5-4.0 (25 °C, 1 M in H2O)

useful pH range

6.5-7.9

pKa (25 °C)

7.2

solubility

H2O: 1 M at 20 °C, clear, colorless

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Al: ≤5 mg/kg
As: ≤0.1 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

λ

1 M in H2O

UV absorption

λ: 260 nm Amax: 0.020
λ: 280 nm Amax: 0.015

SMILES string

OS(=O)(=O)CCCN1CCOCC1

InChI

1S/C7H15NO4S/c9-13(10,11)7-1-2-8-3-5-12-6-4-8/h1-7H2,(H,9,10,11)

InChI key

DVLFYONBTKHTER-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Satisfactory for electron transport studies and phosphorylation in chloroplast lamella preparations; non-toxic buffering system for the extensive growth of Streptomyces clavuligerus and cephalosporin production, in a chemically defined medium; electrolyte system component for the IEF of 2-D gel electrophoresis

Other Notes

Easily compare specifications for MOPS products with the MOPS specification table.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Aharonowitz et al.
Archives of microbiology, 115(2), 169-173 (1977-11-18)
A high concentration of potassium phosphate (75--100 mM) stabilized pH and supported extensive growth of Streptomyces clavuligerus in a chemically defined medium; such a concentration also inhibited cephalosporin production. Although Tris buffer was found to have detrimental effects on growth
J.A.A. Chambers et al.
Electrophoresis, 6, 339-339 (1985)
Junji Suzuki et al.
Nature communications, 5, 4153-4153 (2014-06-14)
The endoplasmic reticulum (ER) and mitochondria accumulate Ca(2+) within their lumens to regulate numerous cell functions. However, determining the dynamics of intraorganellar Ca(2+) has proven to be difficult. Here we describe a family of genetically encoded Ca(2+) indicators, named calcium-measuring
Hydrogen ion buffers for biological research.
N E Good et al.
Biochemistry, 5(2), 467-477 (1966-02-01)
Y S L Lee et al.
Human reproduction (Oxford, England), 30(3), 543-552 (2015-01-09)
What is the relationship between cleavage stage embryo kinetics, blastocyst metabolism and subsequent embryo viability? Embryos cleaving faster at the first cleavage division resulted in blastocysts with a larger inner cell mass (ICM), higher glucose consumption, lower glycolytic rate, higher

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service