Skip to Content
Merck

67627

Sodium chloride 20 mM – HEPES Buffer pH 7.0; 50 mM – tert-Butanol 40% solution

Synonym(s):

Crystallization solution 23/Kit-No 18839

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


pH

7.0±0.2 ( in neat)

density

1.37 g/mL at 20 °C

storage temp.

2-8°C



Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system, Respiratory system

Storage Class

3 - Flammable liquids

Flash Point (°F)

68.0 °F - closed cup

Flash Point (°C)

20 °C - closed cup

PPE (personal protective equipment)

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library