Skip to Content
Merck
All Photos(1)

Key Documents

63259

Sigma-Aldrich

D-myo-Inositol 2,3,4,5,6-pentakis-phosphate

≥90% (HPLC)

Synonym(s):

L-myoInositol 1,2,4,5,6-pentakis-phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H17O21P5
CAS Number:
Molecular Weight:
580.06
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Assay

≥90% (HPLC)

storage temp.

−20°C

SMILES string

O[C@H]1[C@H](OP(O)(O)=O)[C@@H](OP(O)(O)=O)[C@H](OP(O)(O)=O)[C@@H](OP(O)(O)=O)[C@H]1OP(O)(O)=O

InChI

1S/C6H17O21P5/c7-1-2(23-28(8,9)10)4(25-30(14,15)16)6(27-32(20,21)22)5(26-31(17,18)19)3(1)24-29(11,12)13/h1-7H,(H2,8,9,10)(H2,11,12,13)(H2,14,15,16)(H2,17,18,19)(H2,20,21,22)/t1-,2-,3-,4+,5-,6-/m0/s1

InChI key

CTPQAXVNYGZUAJ-CNWJWELYSA-N

Biochem/physiol Actions

Inositides have an important impact on diverse areas of cellular regulation.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christopher J Barker et al.
Methods in molecular biology (Clifton, N.J.), 645, 1-19 (2010-07-21)
Inositides have an important impact on diverse areas of cellular regulation. However, since this area has grown exponentially from the mid 1980s onwards, many workers find themselves relatively new to the field. In this chapter, we establish a broad foundation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service