Select a Size
About This Item
product line
BioUltra
assay
≥99.0% (RT)
impurities
insoluble matter, passes filter test
pH
7.0-8.5 (25 °C, 1 M in H2O)
solubility
H2O: 1 M at 20 °C, clear, colorless
anion traces
chloride (Cl-): ≤50 mg/kg, sulfate (SO42-): ≤50 mg/kg
cation traces
Al: ≤5 mg/kg, As: ≤0.1 mg/kg, Ba: ≤5 mg/kg, Bi: ≤5 mg/kg, Ca: ≤50 mg/kg, Cd: ≤5 mg/kg, Co: ≤5 mg/kg, Cr: ≤5 mg/kg, Cu: ≤5 mg/kg, Fe: ≤5 mg/kg, Li: ≤5 mg/kg, Mg: ≤10 mg/kg, Mn: ≤5 mg/kg, Mo: ≤5 mg/kg, Na: ≤200 mg/kg, Ni: ≤5 mg/kg, Pb: ≤5 mg/kg, Sr: ≤5 mg/kg, Zn: ≤5 mg/kg
absorption
cut-off at 286 nm in H2O at 1 M
SMILES string
O.[K+].[K+].[O-]C(=O)C([O-])=O
InChI
1S/C2H2O4.2K.H2O/c3-1(4)2(5)6;;;/h(H,3,4)(H,5,6);;;1H2/q;2*+1;/p-2
InChI key
QCPTVXCMROGZOL-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Other Notes
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service