Skip to Content
Merck

57490

Inosine-5′-diphosphate trisodium salt

≥80% (HPLC)

Synonym(s):

5′-IDP-Na3

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H11N4Na3O11P2
CAS Number:
Molecular Weight:
494.13
EC Number:
275-821-2
UNSPSC Code:
41106305
PubChem Substance ID:
Beilstein/REAXYS Number:
8818109
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

assay

≥80% (HPLC)

form

crystalline

impurities

≤15% IMP, ≤5% ITP

storage temp.

−20°C

SMILES string

[Na+].[Na+].[Na+].O[C@H]1[C@@H](O)[C@@H](O[C@@H]1COP([O-])(=O)OP([O-])([O-])=O)n2cnc3C(=O)NC=Nc23

InChI

1S/C10H14N4O11P2.3Na/c15-6-4(1-23-27(21,22)25-26(18,19)20)24-10(7(6)16)14-3-13-5-8(14)11-2-12-9(5)17;;;/h2-4,6-7,10,15-16H,1H2,(H,21,22)(H,11,12,17)(H2,18,19,20);;;/q;3*+1/p-3/t4-,6-,7-,10-;;;/m1.../s1

InChI key

CPIQGMJSIPVOOS-MSQVLRTGSA-K

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lizhen Qiao et al.
Journal of chromatography. A, 1360, 240-247 (2014-08-19)
Glucaminium-based ionic liquids are a new class of recently developed ionic liquids and prepared by functionalizing the amine group of N-methyl-d-glucamine, which renders them good hydrophilicity due to the presence of the glucose structure and charged quaternary ammonium group. In

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service