Skip to Content
Merck

51431

Atto Rho11 maleimide

BioReagent, suitable for fluorescence

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.32
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

form

solid

manufacturer/tradename

ATTO-TEC GmbH

fluorescence

λex 671 nm; λem 595 nm

suitability

suitable for fluorescence

storage temp.

−20°C

Application

Atto Rho11 maleimide is a new rhodamine dye featuring a functionality for coupling to bio-molecules such as DNA, RNA, proteins etc. The label shows very high fluorescence efficiency and exceptionally high photostability. The dye is moderately hydrophilic. ATTO Rho11 is a cationic dye. After coupling to a substrate the label carries a net electrical charge of +1. Maleimides are well suited for coupling to thiol groups. This is similar to iodacetamides, but maleimides react more selectively with thiols. They do not show significant reaction with histidine or methionine. Hydrolysis of maleimides to a mixture of isomeric nonreactive maleamic acids can compete significantly with thiol modification, particularly above pH 8. Maleimides may be used for labeling of amines, which usually requires a higher pH than reaction of maleimides with thiols.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library