Skip to Content
Merck

490M-1

Tyrosine Hydroxylase (LNC1) Mouse Monoclonal Antibody

Synonym(s):

TYH

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41116161
Conjugate:
unconjugated
Clone:
LNC1
Application:
IHC
Citations:
4
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

(Mouse)

Quality Segment

conjugate

unconjugated

antibody form

(IgG1 Supernatant)

antibody product type

primary antibodies

clone

LNC1

description

For In Vitro Diagnostic Use in Select Regions

form

buffered aqueous solution

species reactivity

human

packaging

vial of 0.1 mL (concentrate (490M-14)), vial of 0.1 mL (concentrate (490M-14-RUO) Research Use Only), vial of 0.5 mL (concentrate (490M-15)), vial of 1.0 mL (concentrate (490M-16)), vial of 1.0 mL (concentrate (490M-16-RUO) Research Use Only), vial of 1.0 mL (pre-dilute (490M-17-RUO) Research Use Only), vial of 1.0 mL (pre-dilute ready-to-use (490M-17)), vial of 7.0 mL (pre-dilute ready-to-use (490M-18)), vial of 7.0 mL (pre-dilute ready-to-use (490M-18-RUO) Research Use Only)

manufacturer/tradename

Cell Marque®

technique(s)

immunohistochemistry: suitable (formalin-fixed, paraffin-embedded sections 1:25-1:100 (concentrated))

isotype

IgG1

control

neuroblastoma

shipped in

wet ice

storage temp.

2-8°C

visualization

cytoplasmic

General description

Tyrosine Hydroxylase (TYH) is the rate-limiting enzyme of catecholamine biosynthesiss. TYH uses tetrahydrobiopterin and molecular oxygen to convert tyrosine to L-DOPA, a precursor to dopamine. Anti-TYH is sensitive and specific for peripheral neuroblastic tumors in the differential diagnosis from other tumors of childhood and shows high diagnostic utility. Anti-TYH has been shown to positively label all pheochromocytomas and sympathetic paragangliomas, and thus aids in differentiating them from their histologic mimics such as adrenocortical neoplasms.

Physical form

Solution in Tris Buffer, pH 7.3-7.7, with 1% BSA and <0.1% Sodium Azide.

Preparation Note

Download the IFU specific to your product lot and format
Note: This requires a keycode which can be found on your packaging or product label.

Analysis Note

United States - IVD
Canada - RUO
European Union - IVD
Japan - RUO

Other Notes

For Technical Service please contact: 800-665-7284 or email: [email protected]

Legal Information

Cell Marque is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library