Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
Empirical Formula (Hill Notation):
C34H37N3O8S4
CAS Number:
Molecular Weight:
743.93
PubChem Substance ID:
UNSPSC Code:
12352200
fluorescence
λex 588 nm; λem 609 nm in 0.1 M phosphate pH 7.0
suitability
suitable for fluorescence
storage temp.
−20°C
SMILES string
CS(=O)(=O)SCCNS(=O)(=O)c1ccc(c(c1)S([O-])(=O)=O)C2=C3C=C4CCC[N+]5=C4C(CCC5)=C3Oc6c7CCCN8CCCc(cc26)c78
InChI
1S/C34H37N3O8S4/c1-47(38,39)46-17-12-35-48(40,41)23-10-11-24(29(20-23)49(42,43)44)30-27-18-21-6-2-13-36-15-4-8-25(31(21)36)33(27)45-34-26-9-5-16-37-14-3-7-22(32(26)37)19-28(30)34/h10-11,18-20,35H,2-9,12-17H2,1H3
InChI key
DHYGLLUUGIYOPV-UHFFFAOYSA-N
Application
Texas Red-2-sulfonamidoethyl methanethiosulfonate (Texas Red-2-MTS), S-alkylating, may be use to label S-nitrosothiol and sulfhydryl (cysteine) containing compounds such as proteins for detection with fluorescence applications.
Analysis Note
degree of coupling ≥20%
Legal Information
Texas Red is a registered trademark of Life Technologies
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Yi Yang et al.
Proceedings of the National Academy of Sciences of the United States of America, 102(1), 117-122 (2004-12-25)
Protein S-nitrosation represents a recently described form of post-translational modification that is rapid and reversible. However, the analysis of protein S-nitrosation in situ has been difficult because of the absence of specific probes and the instability of cellular protein S-nitrosothiols.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service