Skip to Content
Merck

Skip To

419-GS

Human Marrow Stromal Cell Growth Supplements (50 ml)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
Please contact Customer Service for Availability
CZK 2,370.00

About This Item

NACRES:
NA.71
UNSPSC Code:
12352207

CZK 2,370.00


Please contact Customer Service for Availability

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

shelf life

1 yr

manufacturer/tradename

Cell Applications, Inc

shipped in

dry ice

storage temp.

−20°C

General description

Added to Basal Medium to create Growth Medium

Preparation Note

contained in protocol

Analysis Note

Each lot was tested for the ability to support plating, spreading and proliferation

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
R419-GSCN419-GS417-GS
form

liquid

form

liquid

form

liquid

form

liquid

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

shipped in

dry ice

shipped in

dry ice

shipped in

dry ice

shipped in

wet ice

shelf life

1 yr

shelf life

1 yr

shelf life

1 yr

shelf life

1 yr

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

manufacturer/tradename

Cell Applications, Inc

manufacturer/tradename

Cell Applications, Inc

manufacturer/tradename

Cell Applications, Inc

manufacturer/tradename

-


Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable

wgk

WGK 1



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters