Skip to Content
Merck

39881

UV-VIS detectable IEF Marker solution

pI 10.1, 1 mg/mL in 10 mM NaOH, 0.45 μm filtered

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.26
UNSPSC Code:
41105336
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


sterility

0.45 μm filtered

concentration

1 mg/mL in 10 mM NaOH

storage temp.

2-8°C

Application

Suitable as IEF marker for HPCE and IEF gels.


Still not finding the right product?


Storage Class

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library