Skip to Content
Merck

39369

Azide MegaStokes dye 805

for click labeling

Synonym(s):

1-(3-Azidopropyl)-4-[(1E)-2-[6-(diethylamino)-2-benzofuranyl]ethenyl]quinolinium hexafluorophosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H28F6N5OP
Molecular Weight:
571.50
PubChem Substance ID:
UNSPSC Code:
12352200
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


fluorescence

λex 602 nm; λem 805 nm±10 nm in DMSO

storage temp.

−20°C

SMILES string

CCN(CC)C1=CC=C2C(OC(/C=C/C3=CC=[N+](CCCN=[N+]=[N-])C4=C3C=CC=C4)=C2)=C1.F[P-](F)(F)(F)(F)F

InChI

1S/C26H28N5O.F6P/c1-3-30(4-2)22-12-10-21-18-23(32-26(21)19-22)13-11-20-14-17-31(16-7-15-28-29-27)25-9-6-5-8-24(20)25;1-7(2,3,4,5)6/h5-6,8-14,17-19H,3-4,7,15-16H2,1-2H3;/q+1;-1

InChI key

OTSNEEDVAQPEPS-UHFFFAOYSA-N



pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library