About This Item
UNSPSC Code:
41103801
description
Colder Quick Connect
sterility
sterile; γ-irradiated
packaging
pkg of 10 pkg
General description
Female Quick Cap Polycarbonate for 1/4" and 3/8" Male Quick Connect
Packaging
Individually Bagged
Legal Information
BIOEAZE is a trademark of Sigma-Aldrich Co. LLC
Still not finding the right product?
Explore all of our products under BIOEAZE™ Tubing and Components
Choose from one of the most recent versions:
Already Own This Product?