Skip to Content
Merck
All Photos(1)

Key Documents

31630

Sigma-Aldrich

3,4-Diethoxy-3-cyclobutene-1,2-dione

purum, ≥97.0% (GC)

Synonym(s):

Diethyl squarate, Squaric acid diethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2C4(=O)2
CAS Number:
Molecular Weight:
170.16
Beilstein:
640626
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥97.0% (GC)

refractive index

n20/D 1.509 (lit.)
n20/D 1.509

bp

95 °C/0.1 mmHg (lit.)

density

1.15 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCOC1=C(OCC)C(=O)C1=O

InChI

1S/C8H10O4/c1-3-11-7-5(9)6(10)8(7)12-4-2/h3-4H2,1-2H3

InChI key

DFSFLZCLKYZYRD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Building block for the synthesis of substituted and fused p-benzoquinones and of cyclopentenediones

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S.T. Perri et al.
Journal of the American Chemical Society, 112, 1897-1897 (1990)
S. Iyer et al.
Journal of the American Chemical Society, 109, 2759-2759 (1987)
L.D. Foland et al.
Journal of the American Chemical Society, 111, 975-975 (1989)
M.W. Reed et al.
The Journal of Organic Chemistry, 53, 4166-4166 (1988)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service