Skip to Content
Merck
All Photos(1)

Key Documents

25038

Sigma-Aldrich

2-Chloro-4-methylaniline

≥97.0% (GC)

Synonym(s):

2-Chloro-p-toluidine, 4-Amino-3-chlorotoluene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CH3)NH2
CAS Number:
Molecular Weight:
141.60
Beilstein:
774514
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.0% (GC)

refractive index

n20/D 1.575 (lit.)
n20/D 1.575

bp

223-225 °C (lit.)

mp

7 °C (lit.)

density

1.151 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(N)c(Cl)c1

InChI

1S/C7H8ClN/c1-5-2-3-7(9)6(8)4-5/h2-4H,9H2,1H3

InChI key

XGYLSRFSXKAYCR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

210.2 °F - closed cup

Flash Point(C)

99 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service