Skip to Content
Merck

Skip To

18892

Octyl β-D-1-thiomaltoside

≥99.0% (TLC)

Synonym(s):

Octyl β-D-1-thiomaltopyranoside

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C20H38O10S
CAS Number:
Molecular Weight:
470.57
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12161900
MDL number:


assay

≥99.0% (TLC)

mol wt

470.57 g/mol

impurities

insoluble matter, passes filter test

pH

5-8 (1 % (w/v) in H2O)

λ

1 % (w/v) in H2O

UV absorption

λ: 260 nm Amax: ≤0.20, λ: 280 nm Amax: ≤0.15

SMILES string

CCCCCCCCS[C@@H]1O[C@H](CO)[C@@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O)[C@H](O)[C@H]1O

InChI

1S/C20H38O10S/c1-2-3-4-5-6-7-8-31-20-17(27)15(25)18(12(10-22)29-20)30-19-16(26)14(24)13(23)11(9-21)28-19/h11-27H,2-10H2,1H3/t11-,12-,13-,14+,15-,16-,17-,18-,19-,20+/m1/s1

InChI key

JHBBNAKIOKQRJS-NQXZFOFXSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters