Skip to Content
Merck
All Photos(1)

Key Documents

17784

Sigma-Aldrich

Apiin

≥97.0% (HPLC)

Synonym(s):

Apigenin-7-(2-O-apiosylglucoside)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H28O14
CAS Number:
Molecular Weight:
564.49
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.47

Assay

≥97.0% (HPLC)

form

powder or crystals

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

SMILES string

OC[C@H]1O[C@@H](Oc2cc(O)c3C(=O)C=C(Oc3c2)c4ccc(O)cc4)[C@H](O[C@@H]5OC[C@](O)(CO)[C@@H]5O)[C@@H](O)[C@@H]1O

InChI

1S/C26H28O14/c27-8-18-20(32)21(33)22(40-25-23(34)26(35,9-28)10-36-25)24(39-18)37-13-5-14(30)19-15(31)7-16(38-17(19)6-13)11-1-3-12(29)4-2-11/h1-7,18,20-25,27-30,32-35H,8-10H2/t18-,20-,21+,22-,23-,24-,25+,26-/m1/s1

InChI key

NTDLXWMIWOECHG-MFMSCHSJSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Apiin is one of the major flavonoids found in the aromatic herb parsley. It is a bi-glycosylated form of apigenin.

Biochem/physiol Actions

Apiin is well-known for its antioxidant properties. It can enhance the production of β-apiosidase, an extracellular glycosidase in Aspergillus niger. Apiin also exerts antiplatelet activity.
Apiin, a major flavonoid component of celery, shows antiinflammatory activity mediated through inhibition of nitric oxide synthesis and inhibition of iNOS expression.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Lechtenberg et al.
Die Pharmazie, 62(12), 949-954 (2008-01-25)
Drying process of parsley leaves from Petroselinum crispum L. can influence the sensory qualities and aromatic taste of this herbal product. Beside oven-dried material, freeze-dried parsley is getting increasingly into the market. In the course of a search for analytical
T Mencherini et al.
The Journal of pharmacy and pharmacology, 59(6), 891-897 (2007-07-20)
Flavonoids, natural compounds widely distributed in the plant kingdom, are reported to affect the inflammatory process and to possess anti-inflammatory as well as immunomodulatory activity in-vitro and in-vivo. Since nitric oxide (NO) produced by inducible nitric oxide synthase (iNOS) is
You-Lan Chen et al.
World journal of gastroenterology, 24(13), 1398-1409 (2018-04-11)
To investigate the protective effects of Ampelopsis grossedentata (AMP) on dextran sulfate sodium (DSS)-induced colitis in mice based on systems pharmacology approach. Systems pharmacology approach was used to predict the active ingredients, candidate targets and the efficacy of AMP on
Tae Fujimori et al.
Carbohydrate research, 477, 20-25 (2019-04-02)
UDP-apiose, a donor substrate of apiosyltransferases, is labile because of its intramolecular self-cyclization ability, resulting in the formation of apiofuranosyl-1,2-cyclic phosphate. Therefore, stabilization of UDP-apiose is indispensable for its availability and identifying and characterizing the apiosyltransferases involved in the biosynthesis
J Kasthuri et al.
Colloids and surfaces. B, Biointerfaces, 68(1), 55-60 (2008-11-04)
We report a novel strategy for the biological synthesis of anisotropic gold and quasi-spherical silver nanoparticles by using apiin as the reducing and stabilizing agent. The size and shape of the nanoparticles can be controlled by varying the ratio of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service