Skip to Content

12244

2-Keto-D-glucose

Synonym(s):

D-Glucosone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H10O6
CAS Number:
Molecular Weight:
178.14
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


SMILES string

OC[C@@H](O)[C@@H](O)[C@H](O)C(=O)C=O

InChI

1S/C6H10O6/c7-1-3(9)5(11)6(12)4(10)2-8/h1,4-6,8,10-12H,2H2/t4-,5-,6-/m1/s1

InChI key

DCNMIDLYWOTSGK-HSUXUTPPSA-N



Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library