Skip to Content
Merck

11544

2′-Azido-2′-deoxyuridine

≥98.0% (N)

Synonym(s):

N3-dU

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H11N5O5
CAS Number:
Molecular Weight:
269.21
EC Number:
248-113-6
UNSPSC Code:
41106305
PubChem Substance ID:
Beilstein/REAXYS Number:
4204828
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

biological source

synthetic

assay

≥98.0% (N)

form

powder

mp

149-153 °C

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@H]([C@H](N=[N+]=[N-])[C@@H]1O)N2C=CC(=O)NC2=O

InChI

1S/C9H11N5O5/c10-13-12-6-7(17)4(3-15)19-8(6)14-2-1-5(16)11-9(14)18/h1-2,4,6-8,15,17H,3H2,(H,11,16,18)/t4-,6-,7-,8-/m1/s1

InChI key

MRUKYOQQKHNMFI-XVFCMESISA-N

Application

2′-Azido-2′-deoxyuridine (N3-dU) upon phosphorylation to 2′-azido-2′-deoxyuridine 5′-diphosphate (N3dUDP) becomes a mechanistic inhibitor of bacterial ribonucleoside diphosphate reductase (RDPR). N3-dU has recently been evaluated for its potential use as a sensitive multi-spectroscopic probe for nucleic acids

Storage Class

13 - Non Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Béatrice Roy et al.
Biochemical pharmacology, 68(4), 711-718 (2004-07-28)
In this paper, the enantioselectivity of ribonucleotide reductase (RNR, EC 1.17.4.1), a pivotal enzyme involved in DNA biosynthesis, was studied using the beta-d and beta-l stereoisomers of 2'-azido-2'-deoxynucleosides of uracil and cytosine. The corresponding 5'-diphosphate derivatives in the d-configuration have
S P Salowe et al.
Biochemistry, 26(12), 3408-3416 (1987-06-16)
Ribonucleoside diphosphate reductase (RDPR) from Escherichia coli was completely inactivated by 1 equiv of the mechanism-based inhibitor 2'-azido-2'-deoxyuridine 5'-diphosphate (N3UDP). Incubation of RDPR with [3'-3H]N3UDP resulted in 0.2 mol of 3H released to solvent per mole of enzyme inactivated, indicating
Matthew J Tucker et al.
Physical chemistry chemical physics : PCCP, 13(6), 2237-2241 (2010-12-01)
The vibrations in the azido-, N(3), asymmetric stretching region of 2'-azido-2'-deoxyuridine (N(3)dU) are examined by two-dimensional infrared spectroscopy. In water and tetrahydrofuran (THF), the spectra display a single sharp diagonal peak that shows solvent sensitivity. The frequency-frequency correlation time in
S H Kang et al.
Nucleosides & nucleotides, 17(6), 1077-1088 (1998-08-26)
2'-Azido-2'-deoxyuridine and 2'-azido-2'-deoxycytidine were evaluated for their inhibitory activity against ribonucleotide reductase and for subsequent cell growth inhibition. Their mono- and di-phosphates were synthesized and their inhibitory activities against the reductase were also determined in a permeabilized cell system, along
S Salowe et al.
Biochemistry, 32(47), 12749-12760 (1993-11-30)
Ribonucleotide reductase (RDPR) from Escherichia coli is composed of two subunits, R1 and R2, and catalyzes the conversion of nucleotides to deoxynucleotides. The mechanism of inactivation of RDPR by 2'-azido-2'-deoxynucleoside 5'-diphosphate (N3UDP) has been examined using a variety of isotopically

Global Trade Item Number

SKUGTIN
302309-250MG04061837656385
M072000-C04061822549746
M072000-5G04061838216694
302309-5G04061826665831
302309-1G04061837656378

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service