Skip to Content
Merck

Skip To

08637

Atto 594

suitable for fluorescence, ≥90.0% (HPLC)

Sign In to View Organizational & Contract Pricing.

About This Item

UNSPSC Code:
12171501
NACRES:
NA.32
MDL number:


assay

≥90.0% (HPLC)

form

(Report result)

manufacturer/tradename

ATTO-TEC GmbH

transmittance

254 nm, >90%, 604 nm, >90%

fluorescence

λex 601 nm; λem 627 nm in 0.1 M phosphate pH 7.0

λ

in ethanol (with 0.1% trifluoroacetic acid)

UV absorption

λ: 598-604 nm Amax

suitability

suitable for fluorescence

detection method

fluorometric

storage temp.

−20°C

General description

Atto 594 is a new label for proteins and oligo-nucleotides with high molecular absorption (120.000) and quantum yield (0.85) as well as sufficient Stoke′s shift between excitation and emission maximum. Atto 594 is characterized by a high photostability, is very hydrophilic, and excellent water solubility.

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters