Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
Empirical Formula (Hill Notation):
C8H7NO3
CAS Number:
Molecular Weight:
165.15
UNSPSC Code:
12352200
NACRES:
NA.79
Product Name
5-Pyridoxolactone, ≥98.5% (HPLC and enzymatic)
InChI
1S/C8H7NO3/c1-4-7(10)6-3-12-8(11)5(6)2-9-4/h2,10H,3H2,1H3
InChI key
PPAXBSPBIWBREI-UHFFFAOYSA-N
assay
≥98.5% (HPLC and enzymatic)
98.5-101.5% (T)
form
powder
technique(s)
HPLC: suitable
color
white to light yellow
storage temp.
−20°C
Biochem/physiol Actions
5-Pyridoxolactone is a normal human metabolite of vitamin B6 metabolism.
General description
5-pyridoxolactone is a metabolite of vitamin B6.
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Vitamins (2013)
Determination of 5-pyridoxic acid, 5-pyridoxic acid lactone, and other vitamin B6 compounds by cation-exchange high-performance liquid chromatography.
J D Mahuren et al.
Methods in enzymology, 280, 22-29 (1997-01-01)
Identification of 5-pyridoxic acid and 5-pyridoxic acid lactone as metabolites of vitamin B6 in humans.
Mahuren, J. D., et al.
The Journal of Nutritional Biochemistry, 2, 449-453 (1991)
Junichi Funami et al.
Biochimica et biophysica acta, 1753(2), 234-239 (2005-10-18)
4-Pyridoxolactonase is involved in the degradation pathway for pyridoxine, a free form of vitamin B6. The gene (mlr6805) encoding the putative 4-pyridoxolactonase of nitrogen fixing symbiotic microorganism Mesorhizobium loti MAFF303099 has been identified based on the genome database. The gene
Jun Peng et al.
Analytica chimica acta, 803, 97-105 (2013-11-13)
A large fraction of the known human metabolome belong to organic acids. However, comprehensive profiling of the organic acid sub-metabolome is a major analytical challenge. In this work, we report an improved method for detecting organic acid metabolites. This method
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service