Skip to Content
Merck

Skip To

05676

Atto 594 DOPE

suitable for fluorescence

Synonym(s):

1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine labeled with Atto 594

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 mg
Please contact Customer Service for Availability
25 800,00 CZK

About This Item

NACRES:
NA.32
UNSPSC Code:
12352108

25 800,00 CZK


Please contact Customer Service for Availability

​
Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥90.0% (TLC)

Quality Segment

manufacturer/tradename

ATTO-TEC GmbH

technique(s)

protein detection: suitable

λ

in chloroform/methanol (8:2)

UV absorption

λ: 603.0-609.0 nm Amax

suitability

suitable for fluorescence

storage temp.

−20°C

General description

Atto 594 is a fluorescent label belonging to the class of Rhodamine dyes. The dye is designed for application in the area of life science, e.g. labeling of DNA, RNA or proteins. Characteristic features of the label are strong absorption, high fluorescence quantum yield, high thermal and photo-stability, excellent water solubility, and very little triplet formation. After coupling to a substrate Atto 594 carries a net electrical charge of -1.

Atto-Dye Labeled Phospholipids
Sigma-Aldrich offers a variety of glycero-phospholipids carrying one or two fatty acid groups (lipophilic groups) and a phosphate ester residue (hydrophilic group). They are labeled at the hydrophilic head group. After incorporation of the phospholipid into a membrane the fluorophore is located at the water/lipid interface of the membrane. We currently provide 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine (DPPE), 1,2-dioleoyl-sn-glycero-3-phosphoethanolamine (DOPE), palmitoyl-sn-glycero-phosphoethanolamine (PPE), and 1,2-dimyristoyl-sn-glycero-3-phospho-ethanolamine (DMPE) labeled with Atto-dyes.

find more information here

Legal Information

This product is for Research use only. In case of intended commercialization, please contact the IP-holder (ATTO-TEC GmbH, Germany) for licensing.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
422080515267335
description

suitable for fluorescence

description

BioReagent, suitable for fluorescence

description

suitable for fluorescence

description

suitable for fluorescence

suitability

suitable for fluorescence

suitability

suitable for fluorescence

suitability

suitable for fluorescence

suitability

suitable for fluorescence

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

assay

≥90.0% (TLC)

assay

≥90.0% (HPCE)

assay

≥90.0% (HPLC)

assay

≥90.0% (HPLC)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

manufacturer/tradename

ATTO-TEC GmbH

manufacturer/tradename

ATTO-TEC GmbH

manufacturer/tradename

ATTO-TEC GmbH

manufacturer/tradename

ATTO-TEC GmbH

UV absorption

λ: 603.0-609.0 nm Amax

UV absorption

-

UV absorption

λ: 643-649 nm Amax

UV absorption

λ: 505.0-511.0 nm Amax


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

  1. Can water be added to ATTO 594 DOPE? Will the product still be usable?

    1 answer
    1. The product labeled as 05676 is DOPE labeled with ATTO 594 and uses chloroform and water as the solvent. However, it's important to note that the product should not come in contact with water. Since the product has been exposed to water, the recommended course of action is to use another vial. Unfortunately, there is no known method to salvage the product once it has come in contact with water.

      Helpful?

Reviews

No rating value

Active Filters