Skip to Content
Merck
All Photos(1)

Key Documents

05130

Sigma-Aldrich

L-Alanine

≥99.0% (NT)

Synonym(s):

(S)-2-Aminopropionic acid, L-α-Aminopropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H7NO2
CAS Number:
Molecular Weight:
89.09
Beilstein:
1720248
EC Number:
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

Quality Level

Assay

≥99.0% (NT)

form

powder or crystals

optical activity

[α]20/D +14.0±1°, c = 5% in 5 M HCl

impurities

≤0.3% foreign amino acids

ign. residue

≤0.1% (as SO4)

loss

≤0.1% loss on drying, 110 °C

color

colorless to white

mp

314.5 °C

solubility

H2O: soluble 89.1 g/l at 20 degrees c
diethyl ether: insoluble
ethanol: soluble (cold 80%)

anion traces

chloride (Cl-): ≤50 mg/kg
sulfate (SO42-): ≤50 mg/kg

cation traces

Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
NH4+: ≤100 mg/kg
Na: ≤50 mg/kg
Ni: ≤5 mg/kg
Pb: ≤5 mg/kg
Zn: ≤5 mg/kg

application(s)

detection
peptide synthesis

SMILES string

C[C@H](N)C(O)=O

InChI

1S/C3H7NO2/c1-2(4)3(5)6/h2H,4H2,1H3,(H,5,6)/t2-/m0/s1

InChI key

QNAYBMKLOCPYGJ-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

Application

L-Alanine is used as a standard to determine absolute concentration of a protein within a mixture of other proteins measured with SI traceability.[1]

Biochem/physiol Actions

Alanine is a nonessential amino acid that plays a key role in the glucose-alanine cycle between muscle tissue and the liver. In amino acid-degrading tissues such as muscle, amino groups are pooled as glutamate by transamination reactions. The amino group of glutamate is transferred to pyruvate via alanine aminotransferase, forming alanine and α-ketoglutarate. The alanine is passed into the blood and transported to the liver. This reaction is reversed in the liver where pyruvate can be used in gluconeogenesis to form glucose, which may return to other tissues through the circulatory system. Increased alanine levels correlate with higher blood pressure, energy intake, cholesterol levels, and body mass index.
L-alanine has the ability to promote the expression of ferritin and antioxidant stress proteins, heme oxygenase-1 (HO-1).[2]

Preparation Note

L-Alanine completely dissolves in water at a concentration of 89.1 g/l at 20°C to yield solution. It is also soluble in 80% ethanol and insoluble in ether.

Other Notes

Preparation of 2-chloropropionic acid[3][4]; Reduction of derivatives to the aldehyde[5][6]

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B. Koppenhoefer et al.
Synthesis, 316-316 (1982)
H. Kogen et al.
Journal of the Chemical Society. Chemical Communications, 311-311 (1987)
K. Watanabe et al.
Synthesis, 225-225 (1987)
Nina Grosser et al.
Biochemical and biophysical research communications, 314(2), 351-355 (2004-01-22)
The amino acid L-alanine has been shown to exert long-term cytoprotection by as yet unidentified molecular mechanisms. Using cultured human endothelial cells (ECV 304), the present study investigates the effect of L-alanine on hydrogen peroxide-mediated cytotoxicity and expression of the
M. Narita et al.
Tetrahedron Letters, 23, 525-525 (1982)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service