Skip to Content
Merck

Skip To

040-50HEM

Freezing Medium for HEM (50 ml)

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
1 ea
Please contact Customer Service for Availability
2 200,00 CZK

About This Item

NACRES:
NA.71
UNSPSC Code:
12352207
Form:
liquid
Shipped in:
wet ice

2 200,00 CZK


Please contact Customer Service for Availability

Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

shelf life

6 mo.

shipped in

wet ice

storage temp.

2-8°C

General description

We offer freezing media that provides optimal cell survival/recovery after storage. Note that Human Epidermal Melanocytes require their own specialized freezing medium, while most other cells respond well to our regular freezing medium.

Preparation Note

contained in protocol

Analysis Note

Each lot was tested for the ability to support plating, spreading and proliferation

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
040-50SCM011ES-002
form

liquid

form

liquid

form

-

form

liquid

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage temp.

-

shipped in

wet ice

shipped in

wet ice

shipped in

-

shipped in

-

shelf life

6 mo.

shelf life

6 mo.

shelf life

-

shelf life

-


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters