Skip to Content
Merck
All Photos(1)

Key Documents

32305

Sigma-Aldrich

Barium acetate

puriss. p.a., ACS reagent, ≥99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3COO)2Ba
CAS Number:
Molecular Weight:
255.42
Beilstein:
3693411
EC Number:
MDL number:
UNSPSC Code:
12352107
PubChem Substance ID:

grade

ACS reagent
puriss. p.a.

Assay

≥99%

form

crystalline

impurities

≤0.0005% heavy metals (as Pb)
≤0.025% KMnO4 red. matter (as HCOOH)

pH

7-8.5 (20 °C, 5%)

anion traces

chloride (Cl-): ≤5 mg/kg
nitrate (NO3-): ≤20 mg/kg

cation traces

Ca: ≤20 mg/kg
Fe: ≤5 mg/kg
K: ≤30 mg/kg
Mg: ≤10 mg/kg
Na: ≤50 mg/kg
Sr: ≤2000 mg/kg

SMILES string

CC(=O)O[Ba]OC(C)=O

InChI

1S/2C2H4O2.Ba/c2*1-2(3)4;/h2*1H3,(H,3,4);/q;;+2/p-2

InChI key

ITHZDDVSAWDQPZ-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Barium acetate is the barium salt of acetic acid.

Application

Barium acetate was used in the preparation of 0.89[(Bi0.5Na0.5)TiO3]-0.11[BaTiO3] (BNT-BT0.11) precursor solution, required for the fabrication of BNT-BT0.11 thin films. It may be used in the preparation of aluminium triacetate. It may be used for the electrophoretic separation of acidic glycosaminoglycans.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Piezoelectric BNT-BT0.11 thin films processed by sol-gel technique.
Cernea M, et al.
J. Mater. Sci., 46(17), 5621-5627 (2011)
Analytical and preparative separation of acidic glycosaminoglycans by electrophoresis in barium acetate.
E Wessler
Analytical biochemistry, 26(3), 439-444 (1968-12-01)
Eagleson M.
Concise Encyclopedia Chemistry, 51-51 (1994)
Li-Chan Yang et al.
Carbohydrate polymers, 111, 856-863 (2014-07-20)
Dendrobium tosaense is a medicinal Dendrobium species widely used in traditional medicine. This study demonstrated some structural characterizations and immunomodulatory activity of the water-soluble polysaccharides derived from the stem of D. tosaense (DTP). DTP was fractioned using DEAE-650 M anion-exchange

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service