Skip to Content
Merck
All Photos(1)

Documents

11526

Sigma-Aldrich

Lead(II) oxide

meets analytical specification of DAC, BPC, 99-100.5% (complexometric), yellow

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PbO
CAS Number:
Molecular Weight:
223.20
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:

vapor pressure

10 mmHg ( 0 °C)

Assay

99-100.5% (complexometric)

quality

meets analytical specification of DAC, BPC

impurities

≤0.5% insoluble in acetic acid

loss

≤0.2% loss on ignition, 700 °C

color

yellow

mp

886 °C (lit.)

anion traces

chloride (Cl-): ≤50 mg/kg

cation traces

Fe: ≤50 mg/kg

SMILES string

O=[PbH2]

InChI

1S/O.Pb

InChI key

YEXPOXQUZXUXJW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Lact. - Repr. 1A - STOT RE 1

Target Organs

Central nervous system,Kidney,Blood

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xingwen Lu et al.
Water research, 47(3), 1353-1360 (2013-01-05)
This study quantified the effects of lead stabilization by blending lead-containing sludge into glass-ceramics through sintering with aluminum- and silica-rich precursors. Using lead oxide (PbO) to simulate lead-laden sludge under thermal conditions, the predominant PbAl(2)O(4) phase was found at temperatures
Deepak Kumar Mohanty et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 89, 264-267 (2012-01-24)
Triply ionized thulium/thulium--ytterbium doped/codoped TeO2-Pb3O4 (TPO) glasses have been fabricated by classical quenching method. The upconversion emission spectra in the Tm3+/Tm3+-Yb3+ doped/codoped glasses upon excitation with a diode laser lasing at ∼980 nm has been studied. Effect of the addition
E M Ibragimova et al.
Journal of nanoscience and nanotechnology, 12(11), 8818-8821 (2013-02-21)
Widely used SiO2-PbO glasses can be a good model for studying electron structure of insulator-metal interface with many Si-O-Pb bonds and eutectic Pb-Si mixture. Gamma-radiation of 60Co with an energy of -1.25 MeV generates efficiently electronic and hole structure defects
Hui Zheng et al.
Huan jing ke xue= Huanjing kexue, 33(3), 857-865 (2012-05-26)
Ti-base lead dioxide electrodes (Ti/PbO2) doped with rare earth La, Ce were prepared by the electrode position. The surface morphology and crystal structure of the electrodes was analyzed by scanning electron microscopy (SEM) and X-ray diffraction (XRD), the results showed
Lei Li et al.
Journal of hazardous materials, 203-204, 274-282 (2012-01-03)
As part of contribution for developing a green recycling process of spent lead acid battery, a nanostructural lead oxide was prepared under the present investigation in low temperature calcination of lead citrate powder. The lead citrate, the precursor for preparation

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service