Z801984
Hellma® calibration standard, neutral density glass filter
F4, for testing the photometric accuracy
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
12352200
NACRES:
SB.32
Recommended Products
grade
certified reference material
manufacturer/tradename
Hellma 666-F4
application(s)
food and beverages
petroleum
pharmaceutical
plastics
format
neat
General description
Neutral density glass filters (nominal absorbance: 1.0 at 440; 465; 546,1; 590; 635 nm)
Application
Hellma® calibration standard, neutral density glass filter for checking the photometric accuracy of spectrophotometers in the visible wavelength range.
Features and Benefits
- Accredited to DAkkS Deutsche Akkreditierungsstelle (National accreditation body for the Federal Republic of Germany)
- Traceable to NIST (National Institute of Standards and Technology)
- Assured Quality of the measurement results
- Proven accuracy and reliability
Other Notes
The most current version of Hellma's handling guidelines for calibration standards can be accessed on the Hellma Analytics web site.
Legal Information
Hellma is a registered trademark of Hellma GmbH
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service