Key Documents
Z219673
Sigma-Aldrich
Aldrich® Abderhalden drying apparatus
pkg of 1 ea
Select a Size
1 EA
CZK 32,300.00
CZK 32,300.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EA
CZK 32,300.00
About This Item
UNSPSC Code:
41121800
NACRES:
NB.43
CZK 32,300.00
Please contact Customer Service for Availability
packaging
pkg of 1 ea
General description
The Aldrich Abderhalden drying apparatus has an improved design features a desiccant flask with a removable drying chamber. Multiple drying chambers may be used to dry several samples in series without dismantling and cleaning the main chamber, flask, and condenser assembly. Dried samples, sealed in the drying chamber, may be easily transferred to a dry box for sample removal under inert atmosphere.
Complete unit consists of a drying chamber (25.5 cm L × 25.4 mm I.D.), desiccant flask with a 2 mm glass stopcock, vapor chamber, Allihn condenser, and 250 mL boiling flask.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)