Skip to Content
Merck
All Photos(1)

Key Documents

Y0001925

Betamethasone dipropionate for system suitability A

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Betamethasone 17,21-dipropionate, 9α-Fluoro-16β-methyl-11β,17α,21-trihydroxy-1,4-pregnadiene-3,20-dione 17,21-dipropionate

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
CZK 3,520.00

CZK 3,520.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
CZK 3,520.00

About This Item

Empirical Formula (Hill Notation):
C28H37FO7
CAS Number:
Molecular Weight:
504.59
EC Number:
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

CZK 3,520.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

betamethasone

manufacturer/tradename

EDQM

format

neat

storage temp.

2-8°C

SMILES string

[H][C@@]12C[C@H](C)[C@](OC(=O)CCl)(C(=O)COC(=O)CC)[C@@]1(C)C[C@H](O)C3(F)[C@@]2([H])CCC4=CC(=O)C=C[C@]34C

InChI

1S/C27H34ClFO7/c1-5-22(33)35-14-21(32)27(36-23(34)13-28)15(2)10-19-18-7-6-16-11-17(30)8-9-24(16,3)26(18,29)20(31)12-25(19,27)4/h8-9,11,15,18-20,31H,5-7,10,12-14H2,1-4H3/t15-,18-,19-,20-,24-,25-,26?,27-/m0/s1

InChI key

UOCNBCVPZWLUKX-HOBFILNXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Betamethasone dipropionate for system suitability A EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Biochem/physiol Actions

Betamethasone 17,21-dipropionate is a glucocorticoid with anti-inflammatory and immunosuppressive activity.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B - STOT RE 2

Target Organs

Liver,Kidney,Endocrine system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service