Skip to Content
Merck
All Photos(1)

Key Documents

Y0001733

Permethrin (25:75)

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

Permethrin

Sign Into View Organizational & Contract Pricing

Select a Size

350 MG
CZK 3,520.00

CZK 3,520.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
350 MG
CZK 3,520.00

About This Item

Empirical Formula (Hill Notation):
C21H20Cl2O3
CAS Number:
Molecular Weight:
391.29
Beilstein:
5765325
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

CZK 3,520.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

pharmaceutical primary standard

API family

permethrin

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

CC1(C)C(\C=C(/Cl)Cl)C1C(=O)OCc2cccc(Oc3ccccc3)c2

InChI

1S/C21H20Cl2O3/c1-21(2)17(12-18(22)23)19(21)20(24)25-13-14-7-6-10-16(11-14)26-15-8-4-3-5-9-15/h3-12,17,19H,13H2,1-2H3

InChI key

RLLPVAHGXHCWKJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Permethrin (25:75) EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

related product

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service