Skip to Content
Merck
All Photos(1)

Documents

Y0000478

Granisetron impurity B

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

N-[(1R,3r,5S)-9-Methyl-9-azabicyclo[3.3.1]non-3-yl]-1H-indazole-3-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H22N4O
CAS Number:
Molecular Weight:
298.38
UNSPSC Code:
41116107
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

granisetron

manufacturer/tradename

EDQM

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

InChI

1S/C17H22N4O/c1-21-12-5-4-6-13(21)10-11(9-12)18-17(22)16-14-7-2-3-8-15(14)19-20-16/h2-3,7-8,11-13H,4-6,9-10H2,1H3,(H,18,22)(H,19,20)

InChI key

GHVQAOGYNZNTIA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Granisetron impurity B EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service