Y0000032
Salbutamol impurity I
European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(1RS)-2-[(1,1-Dimethylethyl)amino]-1-[3-(hydroxymethyl)-4-benzyloxyphenyl]ethanol, 2-tert-Butylamino-1-[3-(hydroxymethyl)-4-benzyloxyphenyl]ethanol
Select a Size
Select a Size
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
albuterol, salbutamol
manufacturer/tradename
EDQM
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
SMILES string
N(C(C)(C)C)CC(O)c1cc(c(cc1)OCc2ccccc2)CO
InChI
1S/C20H27NO3/c1-20(2,3)21-12-18(23)16-9-10-19(17(11-16)13-22)24-14-15-7-5-4-6-8-15/h4-11,18,21-23H,12-14H2,1-3H3
InChI key
ICDQPCBDGAHBGG-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
General description
Application
Packaging
Other Notes
related product
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service