Skip to Content
Merck
All Photos(1)

Key Documents

W441330

Sigma-Aldrich

trans-3-Hexenyl acetate

≥95%, stabilized, FG

Sign Into View Organizational & Contract Pricing

Select a Size

W441330-SAMPLE-K
CZK 3,940.00
100 G
CZK 6,600.00
1 KG
CZK 24,600.00

CZK 3,940.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
W441330-SAMPLE-K
CZK 3,940.00
100 G
CZK 6,600.00
1 KG
CZK 24,600.00

About This Item

Empirical Formula (Hill Notation):
C8H14O2
CAS Number:
Molecular Weight:
142.20
FEMA Number:
4413
MDL number:
UNSPSC Code:
12164502
Flavis number:
09.928
NACRES:
NA.21
grade:
FG
Halal
Kosher
food allergen:
no known allergens

CZK 3,940.00


Please contact Customer Service for Availability

Request a Bulk Order

grade

FG
Halal
Kosher

Quality Level

Agency

follows IFRA guidelines
meets purity specifications of JECFA

reg. compliance

EU Regulation 1223/2009
EU Regulation 1334/2008 & 178/2002
FDA 21 CFR 117

Assay

≥95%

contains

synthetic α-tocopherol as stabilizer

Documentation

see Safety & Documentation for available documents

food allergen

no known allergens

fragrance allergen

no known allergens

Organoleptic

banana; green; fruity; pear; rum

SMILES string

O(CC\C=C\CC)C(=O)C

InChI

1S/C8H14O2/c1-3-4-5-6-7-10-8(2)9/h4-5H,3,6-7H2,1-2H3/b5-4+

InChI key

NPFVOOAXDOBMCE-SNAWJCMRSA-N

Other Notes

Download our Flavors and Fragrances Catalog to view our entire product line.
Subscribe to our Newsletter to keep up to date on our latest Flavors and Fragrances offerings.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

134.6 °F

Flash Point(C)

57 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service