Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
1.5 mL | Please contact Customer Service for Availability | CZK 1,320.00 |
About This Item
grade
certified reference material
CofA
current certificate can be downloaded
packaging
pkg of 1.5 mL
manufacturer/tradename
RTC UST-127
concentration
2000 μg/mL in dichloromethane: carbon disulfide (50:50)
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
application(s)
environmental
petroleum
format
matrix material
storage temp.
2-30°C
General description
Application
Other Notes
1 of 1
This Item | ||
|---|---|---|
| technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) HPLC: suitable, gas chromatography (GC): suitable | technique(s) HPLC: suitable, gas chromatography (GC): suitable |
| format matrix material | format matrix material | format matrix material |
| Quality Level 100, 300 | Quality Level 100, 300 | Quality Level 300 |
| grade certified reference material | grade certified reference material | grade certified reference material |
| storage temp. 2-30°C | storage temp. 2-30°C | storage temp. 2-30°C |
| manufacturer/tradename RTC UST-127 | manufacturer/tradename RTC UST-157 | manufacturer/tradename RTC CRM355-100 |
Analyte
- Decane
- Docosane
- Dodecane
- Dotriacontane
- Eicosane
- Hexacosane
- Hexadecane
- Hexatriacontane
- Octacosane
- Octadecane
- Octane
- Octatriacontane
- Tetracontane
- Tetracosane
- Tetradecane
- Tetratriacontane
- Triacontane
signalword
Danger
Hazard Classifications
Carc. 2 - Eye Irrit. 2 - Flam. Liq. 2 - Repr. 2 - Skin Irrit. 2 - STOT RE 1 - STOT SE 3
target_organs
Central nervous system, Peripheral nervous system,Central nervous system,Cardio-vascular system,Eyes
Storage Class
3 - Flammable liquids
wgk
WGK 2
flash_point_f
-22.0 °F - closed cup
flash_point_c
-30 °C - closed cup
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
75-09-2
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.





