Skip to Content
Merck
All Photos(1)

Documents

TDS1500

Supelco

TDS 1500 mg/L Calibration Std

certified reference material

Synonym(s):

TDS 1500 mg/L Calibrant

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

CofA

current certificate can be downloaded

manufacturer/tradename

RTC TDS1500-500

concentration

in H2O

application(s)

environmental

format

matrix material

storage temp.

2-30°C

General description

This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.

Application

TDS 1500 mg/L Calibration Std Calibration Standard for R&D and Analytical use only.

Features and Benefits

  • Packed in High Density Polyethylene (HDPE) bottle with 500ml of solution
  • A certificate of analysis (CoA) is included with the product
  • Traceable to recognized national standards, and units of measurement recognized at the National Institute of Standards and Technology (NIST) and International System of units (SI)
  • Gravimetrically performed dilutions Balance used to prepare the standard is calibrated to ISO 17025 requirements
  • Stable, ready to use solution with long minimum shelf life

Other Notes

Values of analytes vary lot to lot. Some lots may not contain all analytes.

related product

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service