Skip to Content
Merck
All Photos(1)

Key Documents

T3937

Sigma-Aldrich

Tetracaine hydrochloride

meets USP testing specifications

Synonym(s):

4-(Butylamino)benzoic acid 2-(dimethylamino)ethyl ester, Amethocaine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H24N2O2 · HCl
CAS Number:
Molecular Weight:
300.82
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.21

Agency

meets USP testing specifications

Quality Level

application(s)

pharmaceutical (small molecule)

SMILES string

Cl.CCCCNc1ccc(cc1)C(=O)OCCN(C)C

InChI

1S/C15H24N2O2.ClH/c1-4-5-10-16-14-8-6-13(7-9-14)15(18)19-12-11-17(2)3;/h6-9,16H,4-5,10-12H2,1-3H3;1H

InChI key

PPWHTZKZQNXVAE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Topical ophthalmic anesthetic; used for spinal anesthesia

Biochem/physiol Actions

Blocks voltage-sensitive release of Ca2+ from sarcoplasmic reticulum.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zachery Koneval et al.
Epilepsia, 61(9), 2022-2034 (2020-08-07)
Initial identification of new investigational drugs for the treatment of epilepsy is commonly conducted in well-established mouse acute and chronic seizure models: for example, maximal electroshock (MES), 6 Hz, and corneal kindling. Comparison of the median effective dose (ED50) of approved
Patrick E McCleskey et al.
Dermatologic surgery : official publication for American Society for Dermatologic Surgery [et al.], 39(1 Pt 1), 82-91 (2013-01-03)
Few published studies have analyzed serum lidocaine levels and individual patient characteristics affecting metabolism after application of compounded topical anesthetics. To measure serum lidocaine levels during and cutaneous side effects after standardized application of 23% lidocaine/7% tetracaine compounded anesthetic to
K Ruetzler et al.
British journal of anaesthesia, 109(5), 790-796 (2012-07-27)
Arterial catheterization is painful and is associated with patient stress and anxiety. Analgesia is usually provided by subcutaneous injection of local anaesthetic. An alternative is topical anaesthesia, such as Rapydan which is a novel topical anaesthetic patch containing 70 mg
Quan Wen et al.
Cornea, 32(2), 179-184 (2012-08-08)
Our recent tissue cross-linking studies have raised the possibility of using aliphatic β-nitroalcohols (BNAs) for pharmacologic, therapeutic corneal cross-linking. The present study was performed to determine the permeability of BNAs and to explore the use of permeability-enhancing agents. Ex vivo
Gabriel C Gaviola et al.
The Laryngoscope, 123(4), 852-858 (2013-02-14)
Transnasal endoscopy is commonly performed in an outpatient otolaryngology setting. Patients are typically administered a topical anesthetic and decongestant prior to this procedure to alleviate discomfort and improve visualization. There is no consensus on which topical anesthetic is most effective

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service