Skip to Content
Merck
All Photos(3)

Documents

STS0001

Sigma-Aldrich

TERGITOL 15-S-30

greener alternative

Non-ionic surfactant

Synonym(s):

Alcohols, C12-14 secondary, ethoxylated, Secondary alcohol ethoxylate 31EO

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
UNSPSC Code:
12161902
NACRES:
NA.28

Quality Level

grade

laboratory grade

description

non-ionic

form

solid

mol wt

average mol wt 1,520 g/mol by calculation

greener alternative product characteristics

Design for Degradation
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

CMC

558 ppm (25°C)

viscosity

92 cP(50 °C)

transition temp

cloud point >100 °C ((1 wt% actives aq solution))
pour point 39 °C

solubility

water: soluble

density

1.053 g/mL at 55 °C

HLB

17.4

greener alternative category

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Green Chemistry. This product is a biodegradable surfactant and is aligned with the 10th principle of Green Chemistry "Design for Degradation".
Tergitol 15-S-30 is a secondary alcohol ethoxylate with 31 ethylene oxide (EO) units.

Features and Benefits

  • Excellent emulsion stabilizer with low reactor residue
  • Provides freeze/thaw & ionic stability
  • Electrolyte solubility
  • High temperature wetting

Legal Information

TERGITOL is a trademark of The Dow Chemical Company ("Dow") or an affiliated company of Dow

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service