Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|
About This Item
Linear Formula:
(CH3CH2)2O
CAS Number:
Molecular Weight:
74.12
NACRES:
NA.07
UNSPSC Code:
41116107
Beilstein/REAXYS Number:
1696894
MDL number:
eCl@ss:
39021002
Product RTC000077 is not currently sold in your country. Contact Technical Service
Quality Segment
grade
certified reference material, pharmaceutical secondary standard
vapor density
2.6 (vs air)
CofA
current certificate can be downloaded
autoignition temp.
320 °F
expl. lim.
36.5 %
packaging
pkg of 20 mL
refractive index
n20/D 1.3530 (lit.)
bp
34.6 °C (lit.)
mp
−116 °C (lit.)
density
0.706 g/mL at 25 °C (lit.)
application(s)
pharmaceutical small molecule
storage temp.
2-30°C
SMILES string
CCOCC
InChI
1S/C4H10O/c1-3-5-4-2/h3-4H2,1-2H3
InChI key
RTZKZFJDLAIYFH-UHFFFAOYSA-N
General description
Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards
Analysis Note
These secondary standards offer multi-traceability to the USP and EP primary standards, where they are available.
Other Notes
This Certified Reference Material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this CRM can be found on the certificate of analysis.
To see an example of a Certificate of Analysis for this material enter LRAB9776 in the Documents slot below. This is an example certificate only and may not be the lot that you receive.
Still not finding the right product?
Explore all of our products under Diethyl Ether
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 4 Oral - Flam. Liq. 1 - STOT SE 3
target_organs
Central nervous system
supp_hazards
Storage Class
3 - Flammable liquids
wgk
WGK 1
flash_point_f
-40.0 °F
flash_point_c
-40 °C
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

