Skip to Content
Merck
All Photos(3)

Documents

RM11176L

Millipore

Yersinia enterocolitica NCTC 11176 Lenticule® discs

30-120 CFU, reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.24

grade

reference material

Quality Level

form

disc (hydrosoluble)

shelf life

limited shelf life, expiry date on the label

application(s)

environmental
food and beverages

storage temp.

−20°C

suitability

Yersinia spp.

Looking for similar products? Visit Product Comparison Guide

General description

This reference material (RM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025. All information regarding the use of this RM can be found on the certificate of analysis.
Download your certificate of analysis at: http://www.sigma-aldrich.com.
LENTICULE® discs are manufactured by Sigma-Aldrich under licence from UK Health Security Agency.

Other Notes

Please specify if you require a single batch - in the remark field when ordering (only for orders with more than 15 packs)

Legal Information

Lenticule is a registered trademark of The Secretary of State for Health (United Kingdom)

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service